missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4-Bis(2-ethylhexyl)-4H-cyclopenta[2,1-b:3,4-b']dithiophene 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 5 g |
| Synonym | 4,4-Bis(2-ethylhexyl)-4H-cyclopenta[1,2-b:5,4-b′]dithiophene |
| Formula Weight | 402.70 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 4,4-Bis(2-ethylhexyl)-4H-cyclopenta[2,1-b:3,4-b′]dithiophene |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |