missing translation for 'onlineSavingsMsg'
Learn More

4,4-Bis(2-ethylhexyl)-4H-cyclopenta[2,1-b:3,4-b']dithiophene 98.0+%, TCI America™

Catalog No. B41021G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41021G 1 g
B41025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41021G Supplier TCI America Supplier No. B41021G
Only null left

Chemical Identifiers

CAS 365547-20-2
Molecular Formula C25H38S2
Molecular Weight (g/mol) 402.699
InChI Key NUCIQEWGTLOQTR-UHFFFAOYSA-N
PubChem CID 59426893
SMILES CCCCC(CC)CC1(C2=C(C3=C1C=CS3)SC=C2)CC(CC)CCCC

Specifications

Quantity 1 g
Synonym 4,4-Bis(2-ethylhexyl)-4H-cyclopenta[1,2-b:5,4-b′C]dithiophene
Formula Weight 402.70
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4,4-Bis(2-ethylhexyl)-4H-cyclopenta[2,1-b:3,4-b′]dithiophene
TSCA No
Recommended Storage Refrigerator