missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Biphenyldicarboxaldehyde 98.0+%, TCI America™

Catalog No. B28541G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28541G 1 g
B28545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28541G Supplier TCI America Supplier No. B28541G
Only null left

Chemical Identifiers

CAS 66-98-8
Molecular Formula C14H10O2
Molecular Weight (g/mol) 210.232
InChI Key FEHLIYXNTWAEBQ-UHFFFAOYSA-N
Synonym 4,4'-biphenyldicarboxaldehyde, 1,1'-biphenyl-4,4'-dicarbaldehyde, 4,4'-diformylbiphenyl, biphenyl-4,4'-dicarbaldehyde, 4,4'-biphenyldicarboxyaldehyde, 1,1'-biphenyl-4,4'-dicarboxaldehyde, p,p'-diformylbiphenyl, 4-4-formylphenyl benzaldehyde, 4,4'-bis formyl biphenyl
PubChem CID 6200
IUPAC Name 4-(4-formylphenyl)benzaldehyde
SMILES C1=CC(=CC=C1C=O)C2=CC=C(C=C2)C=O

Specifications

Melting Point 146°C
Color White-Yellow
Quantity 1 g
Formula Weight 210.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Biphenyldicarboxaldehyde
RTECSNumber DV2800200
TSCA No
Recommended Storage Refrigerator