accessibility menu, dialog, popup

UserName

OO

CAS RN 66-98-8

IUPAC Name: [1,1'-biphenyl]-4,4'-dicarbaldehyde
Synonyms: [1,1'-biphenyl]-4,4'-dicarbaldehyde
Molecular Weight (g/mol): 210.23
Molecular Formula: C14H10O2
InChi Key: FEHLIYXNTWAEBQ-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(C=C1)C1=CC=C(C=O)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)