missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Biphenyldiboronic Acid 95.0+%, TCI America™

Catalog No. p-7119680
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B24901G 1 g
B24905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B24901G Supplier TCI America Supplier No. B24901G
Only null left

Chemical Identifiers

CAS 4151-80-8
Molecular Formula C12H12B2O4
Molecular Weight (g/mol) 241.84
MDL Number MFCD00151795
InChI Key SLHKDOGTVUCXKX-UHFFFAOYSA-N
Synonym 4,4'-biphenyldiboronic acid, 1,1'-biphenyl-4,4'-diyldiboronic acid, biphenyl-4,4'-diboronic acid, 4-4-boronophenyl phenyl boronic acid, biphenyl-4,4'-diyldiboronic acid, pubchem5324, acmc-1apw8, 4,4'-biphenyldiboric acid, 4,4-biphenyldiboronic acid, 4,4'-diboronobiphenyl
PubChem CID 2734608
IUPAC Name [4'-(dihydroxyboranyl)-[1,1'-biphenyl]-4-yl]boronic acid
SMILES OB(O)C1=CC=C(C=C1)C1=CC=C(C=C1)B(O)O

Specifications

Melting Point 300°C
Color White-Yellow
Quantity 1 g
Formula Weight 241.84
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Biphenyldiboronic Acid