accessibility menu, dialog, popup

UserName

HOBOHBOHHO

CAS RN 4151-80-8

IUPAC Name: [4'-(dihydroxyboranyl)-[1,1'-biphenyl]-4-yl]boronic acid
Synonyms: 4,4'-biphenyldiboronic acid
Molecular Weight (g/mol): 241.84
Molecular Formula: C12H12B2O4
InChi Key: SLHKDOGTVUCXKX-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)C1=CC=C(C=C1)B(O)O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)