missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Azoxydianisole 97.0+%, TCI America™

Catalog No. A05545G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A05545G 5 g
A055425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A05545G Supplier TCI America Supplier No. A05545G
Only null left

Chemical Identifiers

CAS 1562-94-3
Molecular Formula C14H14N2O3
Molecular Weight (g/mol) 258.277
MDL Number MFCD00008400
InChI Key KAEZRSFWWCTVNP-UHFFFAOYSA-N
Synonym 4,4'-azoxyanisole, p-azoxyanisole, 4,4'-azoxydianisole, p-azoxydianisole, p,p'-azoxyanisole, 4,4'-dimethoxyazoxybenzene, p,p'-azoxydianisole, paa liquid crystal, p,p'-dimethoxyazoxybenzene, azoxybenzene, 4,4'-dimethoxy
PubChem CID 15277
IUPAC Name (4-methoxyphenyl)-(4-methoxyphenyl)imino-oxidoazanium
SMILES COC1=CC=C(C=C1)N=[N+](C2=CC=C(C=C2)OC)[O-]

Specifications

Quantity 5 g
Formula Weight 258.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Azoxydianisole
EINECSNumber (3)-2828
TSCA Yes