missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Azobis(4-cyanovaleric acid), 98%, cont. ca 18% water

Catalog No. AAH5961122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5961122 100 g
AAH5961114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5961122 Supplier Thermo Scientific Chemicals Supplier No. H5961122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2638-94-0 | C12H16N4O4 | 280.284 g/mol

4,4'-Azobis(4-cyanovaleric acid) is used as a free radical imitator in polymer synthesis such as polyvinyl chloride, polyacrylonitrile, polyvinyl alcohol and synthetic fibers. It is utilized for both heterogeneous and homogenous free-radical polymerizations, and as an initiator in reversible addition-fragmentation chain transfer polymerization (RAFT). It is also used as a blowing agent for plastics and elastomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Azobis(4-cyanovaleric acid) is used as a free radical imitator in polymer synthesis such as polyvinyl chloride, polyacrylonitrile, polyvinyl alcohol and synthetic fibers. It is utilized for both heterogeneous and homogenous free-radical polymerizations, and as an initiator in reversible addition-fragmentation chain transfer polymerization (RAFT). It is also used as a blowing agent for plastics and elastomers.

Solubility
Soluble in water.

Notes
Light senstive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 2638-94-0
Molecular Formula C12H16N4O4
Molecular Weight (g/mol) 280.284
MDL Number MFCD00002799
InChI Key VFXXTYGQYWRHJP-UHFFFAOYSA-N
Synonym 4,4'-azobis 4-cyanovaleric acid, azobis cyanovaleric acid, 4,4'-diazene-1,2-diyl bis 4-cyanopentanoic acid, 4,4'-azobis 4-cyanopentanoic acid, acva, 4,4-azobis 4-cyanovaleric acid, valeric acid, 4,4'-azobis 4-cyano, pentanoic acid, 4,4'-azobis 4-cyano, 4,4'-azobis 4-cyano valeric acid acva
PubChem CID 92938
IUPAC Name 4-[(4-carboxy-2-cyanobutan-2-yl)diazenyl]-4-cyanopentanoic acid
SMILES CC(CCC(=O)O)(C#N)N=NC(C)(CCC(=O)O)C#N

Specifications

Melting Point 118°C to 125°C (decomposition)
Linear Formula HOCOCH2CH2C(CH3)(CN)N=NC(CH3)(CN)CH2CH2COOH
UN Number UN1325
Beilstein 1729856
Solubility Information Soluble in water.
Quantity 100 g
Formula Weight 280.28
Sensitivity Light sensitive
Percent Purity 98%
Assay contains ≈18% water
Chemical Name or Material 4,4'-Azobis(4-cyanovaleric acid)
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H228-H302
Flammable solid.
Harmful if swallowed.
Precautionary Statement GHS P Statement
P210-P280-P240-P241-P301+P312-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber 220-135-0
RTECSNumber YV6190500
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only