accessibility menu, dialog, popup

UserName

H3C(S)OONN(R)CH3OONN(?)

CAS RN 2638-94-0

IUPAC Name: (4S)-4-{2-[(1R)-3-carboxylato-1-cyano-1-methylpropyl]diazen-1-yl}-4-cyano-4-methylbutanoate
Synonyms: (4S)-4-{2-[(1R)-3-carboxylato-1-cyano-1-methylpropyl]diazen-1-yl}-4-cyano-4-methylbutanoate
Molecular Weight (g/mol): 278.27
Molecular Formula: C12H14N4O4
InChi Key: VFXXTYGQYWRHJP-TXEJJXNPSA-L
SMILES: C[C@@](CCC([O-])=O)(N=N[C@](C)(CCC([O-])=O)C#N)C#N
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)