missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Azo-bis(4-cyanovaleric acid), ∽98%, MP Biomedicals™

$345.63 - $345.63

Chemical Identifiers

CAS 2638-94-0
Molecular Formula C12H16N4O4
Molecular Weight (g/mol) 280.284
MDL Number MFCD00002799
InChI Key VFXXTYGQYWRHJP-UHFFFAOYSA-N
Synonym 4,4'-azobis 4-cyanovaleric acid, azobis cyanovaleric acid, 4,4'-diazene-1,2-diyl bis 4-cyanopentanoic acid, 4,4'-azobis 4-cyanopentanoic acid, acva, 4,4-azobis 4-cyanovaleric acid, valeric acid, 4,4'-azobis 4-cyano, pentanoic acid, 4,4'-azobis 4-cyano, 4,4'-azobis 4-cyano valeric acid acva
PubChem CID 92938
IUPAC Name 4-[(4-carboxy-2-cyanobutan-2-yl)diazenyl]-4-cyanopentanoic acid
SMILES CC(CCC(=O)O)(C#N)N=NC(C)(CCC(=O)O)C#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN15041225
View Documents
MP Biomedicals Inc
0215041225
25 g
Each for $345.63
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2638-94-0
280.284
VFXXTYGQYWRHJP-UHFFFAOYSA-N
92938
CC(CCC(=O)O)(C#N)N=NC(C)(CCC(=O)O)C#N
C12H16N4O4
MFCD00002799
4,4'-azobis 4-cyanovaleric acid, azobis cyanovaleric acid, 4,4'-diazene-1,2-diyl bis 4-cyanopentanoic acid, 4,4'-azobis 4-cyanopentanoic acid, acva, 4,4-azobis 4-cyanovaleric acid, valeric acid, 4,4'-azobis 4-cyano, pentanoic acid, 4,4'-azobis 4-cyano, 4,4'-azobis 4-cyano valeric acid acva
4-[(4-carboxy-2-cyanobutan-2-yl)diazenyl]-4-cyanopentanoic acid

Specifications

2638-94-0
∽ 98%
MFCD00002799
VFXXTYGQYWRHJP-UHFFFAOYSA-N
4-[(4-carboxy-2-cyanobutan-2-yl)diazenyl]-4-cyanopentanoic acid
280.284
280.28
4, 4'-Azo-bis(4-cyanovaleric acid), Polymer synthesis
White
C12H16N4O4
4,4'-azobis 4-cyanovaleric acid, azobis cyanovaleric acid, 4,4'-diazene-1,2-diyl bis 4-cyanopentanoic acid, 4,4'-azobis 4-cyanopentanoic acid, acva, 4,4-azobis 4-cyanovaleric acid, valeric acid, 4,4'-azobis 4-cyano, pentanoic acid, 4,4'-azobis 4-cyano, 4,4'-azobis 4-cyano valeric acid acva
CC(CCC(=O)O)(C#N)N=NC(C)(CCC(=O)O)C#N
25 g
92938
Powder
Videos
Safety and Handling

Safety and Handling

storageNote1 : Protect from light.

Recommended Storage : Store at 4°C.

SDS
missing translation for 'documents'

missing translation for 'documents'