missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-Aminopiperidino)pyridine Dihydrochloride 98.0+%, TCI America™
$134.21 - $400.78
Chemical Identifiers
| CAS | 1169396-92-2 |
|---|---|
| Molecular Formula | C10H17Cl2N3 |
| Molecular Weight (g/mol) | 250.167 |
| MDL Number | MFCD06797043 |
| InChI Key | YIMBYMVSBQILFO-UHFFFAOYSA-N |
| Synonym | 4-Amino-1-(4-pyridyl)piperidine Dihydrochloride |
| PubChem CID | 44630068 |
| IUPAC Name | 1-pyridin-4-ylpiperidin-4-amine;dihydrochloride |
| SMILES | C1CN(CCC1N)C2=CC=NC=C2.Cl.Cl |
Chemical Identifiers
| 1169396-92-2 | |
| 250.167 | |
| YIMBYMVSBQILFO-UHFFFAOYSA-N | |
| 44630068 | |
| C1CN(CCC1N)C2=CC=NC=C2.Cl.Cl |
| C10H17Cl2N3 | |
| MFCD06797043 | |
| 4-Amino-1-(4-pyridyl)piperidine Dihydrochloride | |
| 1-pyridin-4-ylpiperidin-4-amine;dihydrochloride |
Specifications
| 1169396-92-2 | |
| C10H17Cl2N3 | |
| 1 g | |
| YIMBYMVSBQILFO-UHFFFAOYSA-N | |
| 1-pyridin-4-ylpiperidin-4-amine;dihydrochloride | |
| 44630068 | |
| ≥98.0% (HPLC,T) | |
| 4-(4-Aminopiperidino)pyridine Dihydrochloride |
| White-Yellow | |
| MFCD06797043 | |
| 4-Amino-1-(4-pyridyl)piperidine Dihydrochloride | |
| C1CN(CCC1N)C2=CC=NC=C2.Cl.Cl | |
| 250.167 | |
| 250.17 | |
| Crystalline Powder |
Safety and Handling
TSCA : No