accessibility menu, dialog, popup

UserName

CAS RN 1169396-92-2

NNH2NHClHCl

CAS RN 1169396-92-2

IUPAC Name: 1-(pyridin-4-yl)piperidin-4-amine dihydrochloride
Synonyms: 1-(pyridin-4-yl)piperidin-4-amine dihydrochloride
Molecular Weight (g/mol): 250.17
Molecular Formula: C10H17Cl2N3
InChi Key: YIMBYMVSBQILFO-UHFFFAOYSA-N
SMILES: Cl.Cl.NC1CCN(CC1)C1=CC=NC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results