missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Aminophenyl)thiomorpholine 1,1-Dioxide 98.0+%, TCI America™

Catalog No. A16695G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16695G 5 g
A16691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A16695G Supplier TCI America Supplier No. A16695G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 105297-10-7
Molecular Formula C10H14N2O2S
Molecular Weight (g/mol) 226.29
MDL Number MFCD06797046
InChI Key PLQZCPNIYKUNPR-UHFFFAOYSA-N
PubChem CID 26370384
IUPAC Name 4-(4-aminophenyl)-1λ⁶-thiomorpholine-1,1-dione
SMILES NC1=CC=C(C=C1)N1CCS(=O)(=O)CC1

Specifications

Melting Point 194°C
Quantity 5 g
Synonym 4-(1,1-Dioxothiomorpholino)aniline, 4-Thiomorpholinoaniline 1′,1′-Dioxide
Formula Weight 226.29
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Aminophenyl)thiomorpholine 1,1-Dioxide