accessibility menu, dialog, popup

UserName

SOONNH2

CAS RN 105297-10-7

IUPAC Name: 4-(4-aminophenyl)-1λ⁶-thiomorpholine-1,1-dione
Synonyms: 4-(4-aminophenyl)-1λ⁶-thiomorpholine-1,1-dione
Molecular Weight (g/mol): 226.29
Molecular Formula: C10H14N2O2S
InChi Key: PLQZCPNIYKUNPR-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)N1CCS(=O)(=O)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results