missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Amino-3-fluorophenoxy)-N-methyl-2-pyridinecarboxamide 98.0+%, TCI America™

Catalog No. p-7118177
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2979200MG 200 mg
A29791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2979200MG Supplier TCI America Supplier No. A2979200MG

Chemical Identifiers

CAS 757251-39-1
Molecular Formula C13H12FN3O2
Molecular Weight (g/mol) 261.256
MDL Number MFCD09033852
InChI Key ZQHJPIPGBODVTG-UHFFFAOYSA-N
Synonym 4-(4-Amino-3-fluorophenoxy)-N-methylpicolinamide
PubChem CID 46835219
IUPAC Name 4-(4-amino-3-fluorophenoxy)-N-methylpyridine-2-carboxamide
SMILES CNC(=O)C1=NC=CC(=C1)OC2=CC(=C(C=C2)N)F

Specifications

Melting Point 143°C
Color White-Yellow
Quantity 200 mg
Formula Weight 261.26
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Amino-3-fluorophenoxy)-N-methyl-2-pyridinecarboxamide