accessibility menu, dialog, popup

UserName

CH3NHONOH2NF

CAS RN 757251-39-1

IUPAC Name: 4-(4-amino-3-fluorophenoxy)-N-methylpyridine-2-carboxamide
Synonyms: 4-(4-amino-3-fluorophenoxy)-N-methylpyridine-2-carboxamide
Molecular Weight (g/mol): 261.26
Molecular Formula: C13H12FN3O2
InChi Key: ZQHJPIPGBODVTG-UHFFFAOYSA-N
SMILES: CNC(=O)C1=NC=CC(OC2=CC(F)=C(N)C=C2)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results