missing translation for 'onlineSavingsMsg'
Learn More

4,4'-(alpha-Methylbenzylidene)bisphenol 98.0+%, TCI America™

Catalog No. M1098500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1098500G 500 g
M109825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1098500G Supplier TCI America Supplier No. M1098500G
Only null left

Chemical Identifiers

CAS 1571-75-1
Molecular Formula C20H18O2
Molecular Weight (g/mol) 290.362
MDL Number MFCD00134687
InChI Key VOWWYDCFAISREI-UHFFFAOYSA-N
PubChem CID 623849
IUPAC Name 4-[1-(4-hydroxyphenyl)-1-phenylethyl]phenol
SMILES CC(C1=CC=CC=C1)(C2=CC=C(C=C2)O)C3=CC=C(C=C3)O

Specifications

Melting Point 189°C
Color White
Quantity 500 g
Synonym 1,1-Bis(4-hydroxyphenyl)-1-phenylethane, 4,4′-(1-Phenylethylidene)diphenol
Formula Weight 290.36
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-(alpha-Methylbenzylidene)bisphenol
EINECSNumber (4)-1537
TSCA No