missing translation for 'onlineSavingsMsg'
Learn More

4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium Chloride 98.0+%, TCI America™

Catalog No. p-7127564
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29195G 5 g
D291925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D29195G Supplier TCI America Supplier No. D29195G

Chemical Identifiers

CAS 3945-69-5
Molecular Formula C10H17ClN4O3
Molecular Weight (g/mol) 276.72
MDL Number MFCD03613550
InChI Key BMTZEAOGFDXDAD-UHFFFAOYSA-M
Synonym dmtmm, 4-4,6-dimethoxy-1,3,5-triazin-2-yl-4-methylmorpholinium chloride, 4-4,6-dimethoxy-1,3,5-triazin-2-yl-4-methyl morpholinium chloride, dmt-mm, 4-4,6-dimethoxy-1,3,5-triazine-2-yl-4-methyl morpholinium chloride, 4-4,6-dimethoxy-1,3,5-triazin-2-yl-4-methylmorpholin-4-ium chloride, kunishima coupling reagent, mmtm, 4-4,6-dimethoxy-1,3,5 triazin-2-yl-4-methyl-morpholin-4-ium chloride
PubChem CID 2734059
IUPAC Name 4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholin-4-ium chloride
SMILES [Cl-].COC1=NC(=NC(OC)=N1)[N+]1(C)CCOCC1

Specifications

Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 276.72
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium Chloride
TSCA No
Recommended Storage Freezer