accessibility menu, dialog, popup

UserName

CH3ONOCH3NNNH3COCl

CAS RN 3945-69-5

IUPAC Name: 4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholin-4-ium chloride
Synonyms: dmtmm
Molecular Weight (g/mol): 276.72
Molecular Formula: C10H17ClN4O3
InChi Key: BMTZEAOGFDXDAD-UHFFFAOYSA-M
SMILES: [Cl-].COC1=NC(=NC(OC)=N1)[N+]1(C)CCOCC1
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)