missing translation for 'onlineSavingsMsg'
Learn More

4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride hydrate, 97+%

CAS: 3945-69-5 | C10H17ClN4O3 | 276.72 g/mol

$404.26 - $404.26

Chemical Identifiers

CAS 3945-69-5
Molecular Formula C10H17ClN4O3
Molecular Weight (g/mol) 276.72
MDL Number MFCD03613550
InChI Key BMTZEAOGFDXDAD-UHFFFAOYSA-M
Synonym dmtmm, 4-4,6-dimethoxy-1,3,5-triazin-2-yl-4-methylmorpholinium chloride, 4-4,6-dimethoxy-1,3,5-triazin-2-yl-4-methyl morpholinium chloride, dmt-mm, 4-4,6-dimethoxy-1,3,5-triazine-2-yl-4-methyl morpholinium chloride, 4-4,6-dimethoxy-1,3,5-triazin-2-yl-4-methylmorpholin-4-ium chloride, kunishima coupling reagent, mmtm, 4-4,6-dimethoxy-1,3,5 triazin-2-yl-4-methyl-morpholin-4-ium chloride
PubChem CID 2734059
IUPAC Name 4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholin-4-ium;chloride
SMILES [Cl-].COC1=NC(=NC(OC)=N1)[N+]1(C)CCOCC1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2633306
View Documents
Thermo Scientific Chemicals
H2633306
5 g
Each for $404.26
Only null left
 
Description

Description

4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride hydrate is used as a pharmaceutical intermediate. It is used as a reagent for activating carboxylic acids in solution and solid phase peptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride hydrate is used as a pharmaceutical intermediate. It is used as a reagent for activating carboxylic acids in solution and solid phase peptide synthesis.

Solubility
Soluble in water, Methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. Store under dry inert gas. It is sensitive to air.
Specifications

Chemical Identifiers

3945-69-5
276.72
BMTZEAOGFDXDAD-UHFFFAOYSA-M
2734059
[Cl-].COC1=NC(=NC(OC)=N1)[N+]1(C)CCOCC1
C10H17ClN4O3
MFCD03613550
dmtmm, 4-4,6-dimethoxy-1,3,5-triazin-2-yl-4-methylmorpholinium chloride, 4-4,6-dimethoxy-1,3,5-triazin-2-yl-4-methyl morpholinium chloride, dmt-mm, 4-4,6-dimethoxy-1,3,5-triazine-2-yl-4-methyl morpholinium chloride, 4-4,6-dimethoxy-1,3,5-triazin-2-yl-4-methylmorpholin-4-ium chloride, kunishima coupling reagent, mmtm, 4-4,6-dimethoxy-1,3,5 triazin-2-yl-4-methyl-morpholin-4-ium chloride
4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholin-4-ium;chloride

Specifications

3945-69-5
C10H17ClN4O3
5 g
8026872
Soluble in water,Methanol.
[Cl-].COC1=NC(=NC(OC)=N1)[N+]1(C)CCOCC1
276.72
276.72 (Anhydrous)
4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride hydrate
110°C to 114°C
MFCD03613550
UN3261
dmtmm, 4-4,6-dimethoxy-1,3,5-triazin-2-yl-4-methylmorpholinium chloride, 4-4,6-dimethoxy-1,3,5-triazin-2-yl-4-methyl morpholinium chloride, dmt-mm, 4-4,6-dimethoxy-1,3,5-triazine-2-yl-4-methyl morpholinium chloride, 4-4,6-dimethoxy-1,3,5-triazin-2-yl-4-methylmorpholin-4-ium chloride, kunishima coupling reagent, mmtm, 4-4,6-dimethoxy-1,3,5 triazin-2-yl-4-methyl-morpholin-4-ium chloride
BMTZEAOGFDXDAD-UHFFFAOYSA-M
4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholin-4-ium;chloride
2734059
≥97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P363-P501c

H302-H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only