missing translation for 'onlineSavingsMsg'
Learn More

4-(4,5-Diphenyl-1H-imidazol-2-yl)benzoic Acid 98.0+%, TCI America™

Catalog No. D28165G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D28165G Supplier TCI America Supplier No. D28165G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5496-35-5
Molecular Formula C22H16N2O2
Molecular Weight (g/mol) 340.382
MDL Number MFCD00999654
InChI Key BCXPNUSETAZHEQ-UHFFFAOYSA-N
PubChem CID 192811
IUPAC Name 4-(4,5-diphenyl-1H-imidazol-2-yl)benzoic acid
SMILES C1=CC=C(C=C1)C2=C(N=C(N2)C3=CC=C(C=C3)C(=O)O)C4=CC=CC=C4

Specifications

Color Yellow
Quantity 5 g
Formula Weight 340.38
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4,5-Diphenyl-1H-imidazol-2-yl)benzoic Acid