accessibility menu, dialog, popup

UserName

HOONHN

CAS RN 5496-35-5

IUPAC Name: 4-(4,5-diphenyl-1H-imidazol-2-yl)benzoic acid
Synonyms: 4-(4,5-diphenyl-1H-imidazol-2-yl)benzoic acid
Molecular Weight (g/mol): 340.38
Molecular Formula: C22H16N2O2
InChi Key: BCXPNUSETAZHEQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C1=NC(=C(N1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results