missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-(naphthalen-1-ylmethyl)-1,3,2-dioxaborolane 95.0+%, TCI America™

Catalog No. T36261G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T36261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T36261G Supplier TCI America Supplier No. T36261G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 475250-57-8
Molecular Formula C17H21BO2
Molecular Weight (g/mol) 268.163
MDL Number MFCD22419278
InChI Key VYMBZQMGIGIALJ-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-naphthalen-1-ylmethyl-1,3,2-dioxaborolane, naphthalen-1-ylmethyl boronic acid pinacol ester, 2-1-naphthylmethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl methyl naphthalene
PubChem CID 11254165
IUPAC Name 4,4,5,5-tetramethyl-2-(naphthalen-1-ylmethyl)-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CC2=CC=CC3=CC=CC=C32

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 268.16
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4,5,5-Tetramethyl-2-(naphthalen-1-ylmethyl)-1,3,2-dioxaborolane
TSCA No
Recommended Storage Refrigerator