accessibility menu, dialog, popup

UserName

CAS RN 475250-57-8

IUPAC Name: 4,4,5,5-tetramethyl-2-[(naphthalen-1-yl)methyl]-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(naphthalen-1-ylmethyl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 268.16
Molecular Formula: C17H21BO2
InChi Key: VYMBZQMGIGIALJ-UHFFFAOYSA-N
SMILES: CC1(C)OB(CC2=C3C=CC=CC3=CC=C2)OC1(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results