missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-[3-(triphenylen-2-yl)phenyl]-1,3,2-dioxaborolane 98.0+%, TCI America™

Catalog No. T3261200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3261200MG 200 mg
T32611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3261200MG Supplier TCI America Supplier No. T3261200MG

Chemical Identifiers

CAS 1115639-92-3
Molecular Formula C30H27BO2
Molecular Weight (g/mol) 430.35
MDL Number MFCD28130383
InChI Key NZTBACVVLACPNL-UHFFFAOYSA-N
Synonym 2-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]triphenylene
PubChem CID 58489416
IUPAC Name 4,4,5,5-tetramethyl-2-[3-(triphenylen-2-yl)phenyl]-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)C1=CC=C2C3=CC=CC=C3C3=CC=CC=C3C2=C1

Specifications

Melting Point 179°C
Color White-Yellow
Quantity 200 mg
Formula Weight 430.35
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4,5,5-Tetramethyl-2-[3-(triphenylen-2-yl)phenyl]-1,3,2-dioxaborolane