missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-[3-(trifluoromethoxy)phenyl]-1,3,2-dioxaborolane 98.0+%, TCI America™

Catalog No. T33771G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33771G 1 g
T33775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33771G Supplier TCI America Supplier No. T33771G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 262376-31-8
Molecular Formula C13H16BF3O3
Molecular Weight (g/mol) 288.07
MDL Number MFCD05863919
InChI Key BUSBFOTXIPZTDH-UHFFFAOYSA-N
Synonym 3-(Trifluoromethoxy)phenylboronic Acid Pinacol Ester, 1-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethoxy)benzene
PubChem CID 2760602
IUPAC Name 4,4,5,5-tetramethyl-2-[3-(trifluoromethoxy)phenyl]-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC(OC(F)(F)F)=CC=C1

Specifications

Color Colorless
Quantity 1 g
Formula Weight 288.07
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4,4,5,5-Tetramethyl-2-[3-(trifluoromethoxy)phenyl]-1,3,2-dioxaborolane