accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3COFFF

CAS RN 262376-31-8

IUPAC Name: 4,4,5,5-tetramethyl-2-[3-(trifluoromethoxy)phenyl]-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-[3-(trifluoromethoxy)phenyl]-1,3,2-dioxaborolane
Molecular Weight (g/mol): 288.07
Molecular Formula: C13H16BF3O3
InChi Key: BUSBFOTXIPZTDH-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(OC(F)(F)F)=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results