missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-(2-nitrophenyl)-1,3,2-dioxaborolane 98.0+%, TCI America™

Catalog No. p-7134077
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N10061G 1 g
N10065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N10061G Supplier TCI America Supplier No. N10061G
Only null left

Chemical Identifiers

CAS 190788-59-1
Molecular Formula C12H16BNO4
Molecular Weight (g/mol) 249.07
MDL Number MFCD02179447
InChI Key VLJYUDGCEKORNG-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-2-nitrophenyl-1,3,2-dioxaborolane, 2-nitrobenzeneboronic acid pinacol ester, 2-nitrophenyl boronic acid pinacol ester, 2-nitrophenylboronic acid pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-nitrophenyl, 2-2-nitrophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nitrobenzene, amtb916, 2-nitrophenyl boronic acid, pinacol ester, 2-nitrophenylboronic acid, pinacol ester
PubChem CID 3699568
IUPAC Name 4,4,5,5-tetramethyl-2-(2-nitrophenyl)-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 47°C
Color White-Yellow
Quantity 1 g
Formula Weight 249.07
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4,5,5-Tetramethyl-2-(2-nitrophenyl)-1,3,2-dioxaborolane