accessibility menu, dialog, popup

UserName

CAS RN 190788-59-1

IUPAC Name: 4,4,5,5-tetramethyl-2-(2-nitrophenyl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(2-nitrophenyl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 249.07
Molecular Formula: C12H16BNO4
InChi Key: VLJYUDGCEKORNG-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=CC=C1[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)