missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4,5,5-Tetramethyl-2-(2-methyl-1-propenyl)-1,3,2-dioxaborolane 98.0+%, TCI America™
$109.98 - $329.94
Chemical Identifiers
| CAS | 126689-00-7 |
|---|---|
| Molecular Formula | C10H19BO2 |
| Molecular Weight (g/mol) | 182.07 |
| MDL Number | MFCD09842762 |
| InChI Key | LWXHOCHDERDUID-UHFFFAOYSA-N |
| Synonym | (2-Methyl-1-propenyl)boronic Acid Pinacol Ester |
| PubChem CID | 15148447 |
| IUPAC Name | 4,4,5,5-tetramethyl-2-(2-methylprop-1-enyl)-1,3,2-dioxaborolane |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C=C(C)C |
Chemical Identifiers
| 126689-00-7 | |
| 182.07 | |
| LWXHOCHDERDUID-UHFFFAOYSA-N | |
| 15148447 | |
| B1(OC(C(O1)(C)C)(C)C)C=C(C)C |
| C10H19BO2 | |
| MFCD09842762 | |
| (2-Methyl-1-propenyl)boronic Acid Pinacol Ester | |
| 4,4,5,5-tetramethyl-2-(2-methylprop-1-enyl)-1,3,2-dioxaborolane |
Specifications
| 126689-00-7 | |
| 71°C | |
| MFCD09842762 | |
| (2-Methyl-1-propenyl)boronic Acid Pinacol Ester | |
| B1(OC(C(O1)(C)C)(C)C)C=C(C)C | |
| 182.07 | |
| 182.07 | |
| Liquid |
| Yellow | |
| C10H19BO2 | |
| 1 g | |
| LWXHOCHDERDUID-UHFFFAOYSA-N | |
| 4,4,5,5-tetramethyl-2-(2-methylprop-1-enyl)-1,3,2-dioxaborolane | |
| 15148447 | |
| ≥98.0% (GC) | |
| 4,4,5,5-Tetramethyl-2-(2-methyl-1-propenyl)-1,3,2-dioxaborolane |
Safety and Handling
TSCA : No