CAS RN 126689-00-7
CAS RN 126689-00-7
IUPAC Name:
4,4,5,5-tetramethyl-2-(2-methylprop-1-en-1-yl)-1,3,2-dioxaborolane
Synonyms:
4,4,5,5-tetramethyl-2-(2-methylprop-1-en-1-yl)-1,3,2-dioxaborolane
Molecular Weight (g/mol):
182.07
Molecular Formula:
C10H19BO2
InChi Key:
LWXHOCHDERDUID-UHFFFAOYSA-N
SMILES:
CC(C)=CB1OC(C)(C)C(C)(C)O1
1
–
1
of
1
results
1
4,4,5,5-Tetramethyl-2-(2-methyl-1-propenyl)-1,3,2-dioxaborolane 98.0+%, TCI America™
CAS: 126689-00-7 Molecular Formula: C10H19BO2 Molecular Weight (g/mol): 182.07 MDL Number: MFCD09842762 InChI Key: LWXHOCHDERDUID-UHFFFAOYSA-N Synonym: (2-Methyl-1-propenyl)boronic Acid Pinacol Ester PubChem CID: 15148447 IUPAC Name: 4,4,5,5-tetramethyl-2-(2-methylprop-1-enyl)-1,3,2-dioxaborolane SMILES: B1(OC(C(O1)(C)C)(C)C)C=C(C)C