missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-[10-(1-naphthyl)anthracen-9-yl]-1,3,2-dioxaborolane 97.0+%, TCI America™

Catalog No. T33061G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33061G 1 g
T33065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33061G Supplier TCI America Supplier No. T33061G

Chemical Identifiers

CAS 1149804-35-2
Molecular Formula C30H27BO2
Molecular Weight (g/mol) 430.354
InChI Key HZMCBFYBIMINFR-UHFFFAOYSA-N
Synonym 9-(1-Naphthyl)-10-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anthracene, 10-(1-Naphthyl)anthracene-9-boronic Acid Pinacol Ester
PubChem CID 58050304
IUPAC Name 4,4,5,5-tetramethyl-2-(10-naphthalen-1-ylanthracen-9-yl)-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C3C=CC=CC3=C(C4=CC=CC=C24)C5=CC=CC6=CC=CC=C65

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 430.35
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4,5,5-Tetramethyl-2-[10-(1-naphthyl)anthracen-9-yl]-1,3,2-dioxaborolane