missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-(1-naphthyl)-1,3,2-dioxaborolane 98.0+%, TCI America™

Catalog No. p-7137641
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
T32181G 1 g
T32185G 5 g
2 options

Catalog No. T32181G

Supplier: TCI America T32181G

Only null left

Chemical Identifiers

CAS 68716-52-9
Molecular Formula C16H19BO2
Molecular Weight (g/mol) 254.14
MDL Number MFCD05663869
InChI Key WQGRAXGAXSNSDL-UHFFFAOYSA-N
Synonym Naphthalene-1-boronic Acid Pinacol Ester, 1-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene
PubChem CID 2760174
IUPAC Name 4,4,5,5-tetramethyl-2-(naphthalen-1-yl)-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 57°C
Color White
Quantity 1 g
Formula Weight 254.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4,5,5-Tetramethyl-2-(1-naphthyl)-1,3,2-dioxaborolane