CAS RN 68716-52-9
IUPAC Name:
4,4,5,5-tetramethyl-2-(naphthalen-1-yl)-1,3,2-dioxaborolane
Synonyms:
4,4,5,5-tetramethyl-2-(naphthalen-1-yl)-1,3,2-dioxaborolane
Molecular Weight (g/mol):
254.14
Molecular Formula:
C16H19BO2
InChi Key:
WQGRAXGAXSNSDL-UHFFFAOYSA-N
SMILES:
CC1(C)OB(OC1(C)C)C1=C2C=CC=CC2=CC=C1
1
–
1
of
1
results
1
4,4,5,5-Tetramethyl-2-(1-naphthyl)-1,3,2-dioxaborolane 98.0+%, TCI America™
CAS: 68716-52-9 Molecular Formula: C16H19BO2 Molecular Weight (g/mol): 254.14 MDL Number: MFCD05663869 InChI Key: WQGRAXGAXSNSDL-UHFFFAOYSA-N Synonym: Naphthalene-1-boronic Acid Pinacol Ester, 1-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene PubChem CID: 2760174 IUPAC Name: 4,4,5,5-tetramethyl-2-(naphthalen-1-yl)-1,3,2-dioxaborolane SMILES: CC1(C)OB(OC1(C)C)C1=C2C=CC=CC2=CC=C1