missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-(1-methylpiperid-4-yl)-1,3,2-dioxaborolane, 97%

Catalog No. AC431210010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC431210010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC431210010 Supplier Thermo Scientific Chemicals Supplier No. 431210010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1264198-72-2 | C12H24BNO2 | 225.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1264198-72-2
Molecular Formula C12H24BNO2
Molecular Weight (g/mol) 225.14
MDL Number MFCD11506064
InChI Key IMLCYAZBMGOKQS-UHFFFAOYSA-N
Synonym 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl piperidine, 1-methylpiperidine-4-boronic acid pinacol ester, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl piperidine, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl piperidine
PubChem CID 45790007
IUPAC Name 1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)piperidine
SMILES CN1CCC(CC1)B1OC(C)(C)C(C)(C)O1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 25.0°C to 27.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 225.14
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 4,4,5,5-Tetramethyl-2-(1-methylpiperid-4-yl)-1,3,2-dioxaborolane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only