accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CNCH3

CAS RN 1264198-72-2

IUPAC Name: 1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)piperidine
Synonyms: 1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)piperidine
Molecular Weight (g/mol): 225.14
Molecular Formula: C12H24BNO2
InChi Key: IMLCYAZBMGOKQS-UHFFFAOYSA-N
SMILES: CN1CCC(CC1)B1OC(C)(C)C(C)(C)O1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)