missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4,5,5-Tetramethyl-1,3,2-dioxaborolane 97.0+%, TCI America™
Supplier: TCI America T257225G
Specifications
| 4,4,5,5-Tetramethyl-1,3,2-dioxaborolane | |
| Yellow | |
| C6H13BO2 | |
| 25 g | |
| pinacolborane, 4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl, 4,4,5,5-tetramethyl-1,3,2-dioxa-borolane, pinb-te, pubchem12688, acmc-209ggf, pinacolborane 5g, ksc489k6t, ucfsyhmckwnkah-uhfffaoysa-n | |
| CC1(C)OBOC1(C)C | |
| 127.98 | |
| 127.98 | |
| Liquid |
| 25015-63-8 | |
| 43°C | |
| MFCD00674030 | |
| 3399 | |
| UCFSYHMCKWNKAH-UHFFFAOYSA-N | |
| 4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| 6364989 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 25015-63-8 | |
| 127.98 | |
| UCFSYHMCKWNKAH-UHFFFAOYSA-N | |
| 6364989 | |
| CC1(C)OBOC1(C)C |
| C6H13BO2 | |
| MFCD00674030 | |
| pinacolborane, 4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl, 4,4,5,5-tetramethyl-1,3,2-dioxa-borolane, pinb-te, pubchem12688, acmc-209ggf, pinacolborane 5g, ksc489k6t, ucfsyhmckwnkah-uhfffaoysa-n | |
| 4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Safety and Handling
TSCA : No