missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-1,3,2-dioxaborolane 97.0+%, TCI America™

Catalog No. p-7137289
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25725G 5 g
T257225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25725G Supplier TCI America Supplier No. T25725G
Only null left

Chemical Identifiers

CAS 25015-63-8
Molecular Formula C6H13BO2
Molecular Weight (g/mol) 127.98
MDL Number MFCD00674030
InChI Key UCFSYHMCKWNKAH-UHFFFAOYSA-N
Synonym pinacolborane, 4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl, 4,4,5,5-tetramethyl-1,3,2-dioxa-borolane, pinb-te, pubchem12688, acmc-209ggf, pinacolborane 5g, ksc489k6t, ucfsyhmckwnkah-uhfffaoysa-n
PubChem CID 6364989
IUPAC Name 4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OBOC1(C)C

Specifications

Color Yellow
Boiling Point 43°C
Quantity 5 g
UN Number 3399
Formula Weight 127.98
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4,4,5,5-Tetramethyl-1,3,2-dioxaborolane