missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5,5-Pentafluoropentanol, 94%

CAS: 148043-73-6 | C5H7F5O | 178.102 g/mol

Supplier:  Thermo Scientific Chemicals B2151209

 View more versions of this product

Catalog No. AAB2151209


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4,4,5,5,5-Pentafluoropentanol
1.35
63°C (145°F)
CF3CF2(CH2)3OH
MFCD00153224
4,4,5,5,5-pentafluoro-1-pentanol, 4,4,5,5,5-pentafluoropentanol, pentafluoropentanol, 1-pentanol, 4,4,5,5,5-pentafluoro, fulvestrant intermediate, acmc-1bz8x, 3-perfluoroethyl propan-1-ol, 4,4,5,5,5-pentafluoro pentanol, 4,4,5,5,5-pentafluoro-pentanol, 4,4,5,5,5-pentafluorpentan-1-ol
C(CC(C(F)(F)F)(F)F)CO
178.102
178.1
148043-73-6
135°C
C5H7F5O
1.334
10 g
QROUUECTKRZFHF-UHFFFAOYSA-N
4,4,5,5,5-pentafluoropentan-1-ol
547967
94%

Chemical Identifiers

148043-73-6
178.102
QROUUECTKRZFHF-UHFFFAOYSA-N
547967
C(CC(C(F)(F)F)(F)F)CO
C5H7F5O
MFCD00153224
4,4,5,5,5-pentafluoro-1-pentanol, 4,4,5,5,5-pentafluoropentanol, pentafluoropentanol, 1-pentanol, 4,4,5,5,5-pentafluoro, fulvestrant intermediate, acmc-1bz8x, 3-perfluoroethyl propan-1-ol, 4,4,5,5,5-pentafluoro pentanol, 4,4,5,5,5-pentafluoro-pentanol, 4,4,5,5,5-pentafluorpentan-1-ol
4,4,5,5,5-pentafluoropentan-1-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.

P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c

H227-H302

EINECSNumber : 421-360-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only