missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5,5-Pentafluoropentan-1-ol, 95%

Catalog No. AC438430050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC438430050 5 g
AC438430250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC438430050 Supplier Thermo Scientific Chemicals Supplier No. 438430050

CAS: 148043-73-6 | C5H7F5O | 178.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 148043-73-6
Molecular Formula C5H7F5O
Molecular Weight (g/mol) 178.1
InChI Key QROUUECTKRZFHF-UHFFFAOYSA-N
Synonym 4,4,5,5,5-pentafluoro-1-pentanol, 4,4,5,5,5-pentafluoropentanol, pentafluoropentanol, 1-pentanol, 4,4,5,5,5-pentafluoro, fulvestrant intermediate, acmc-1bz8x, 3-perfluoroethyl propan-1-ol, 4,4,5,5,5-pentafluoro pentanol, 4,4,5,5,5-pentafluoro-pentanol, 4,4,5,5,5-pentafluorpentan-1-ol
PubChem CID 547967
IUPAC Name 4,4,5,5,5-pentafluoropentan-1-ol
SMILES C(CC(C(F)(F)F)(F)F)CO

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Boiling Point 134°C to 135°C
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Quantity 5 g
Formula Weight 178.1
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4,4,5,5,5-Pentafluoropentan-1-ol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Avoid release to the environment.
Wash face, hands and any exposed skin thoroughly after handling.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only