missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4,5,5,5-Pentafluoro-1-pentanol 93.0+%, TCI America™
Supplier: TCI America P134225G
Specifications
| 4,4,5,5,5-Pentafluoro-1-pentanol | |
| Yellow | |
| C5H7F5O | |
| 25 g | |
| QROUUECTKRZFHF-UHFFFAOYSA-N | |
| 4,4,5,5,5-pentafluoropentan-1-ol | |
| 547967 | |
| ≥93.0% (GC) |
| 148043-73-6 | |
| 135°C | |
| MFCD00153224 | |
| 4,4,5,5,5-pentafluoro-1-pentanol, 4,4,5,5,5-pentafluoropentanol, pentafluoropentanol, 1-pentanol, 4,4,5,5,5-pentafluoro, fulvestrant intermediate, acmc-1bz8x, 3-perfluoroethyl propan-1-ol, 4,4,5,5,5-pentafluoro pentanol, 4,4,5,5,5-pentafluoro-pentanol, 4,4,5,5,5-pentafluorpentan-1-ol | |
| C(CC(C(F)(F)F)(F)F)CO | |
| 178.102 | |
| 178.10 | |
| Liquid |
Chemical Identifiers
| 148043-73-6 | |
| 178.102 | |
| QROUUECTKRZFHF-UHFFFAOYSA-N | |
| 547967 | |
| C(CC(C(F)(F)F)(F)F)CO |
| C5H7F5O | |
| MFCD00153224 | |
| 4,4,5,5,5-pentafluoro-1-pentanol, 4,4,5,5,5-pentafluoropentanol, pentafluoropentanol, 1-pentanol, 4,4,5,5,5-pentafluoro, fulvestrant intermediate, acmc-1bz8x, 3-perfluoroethyl propan-1-ol, 4,4,5,5,5-pentafluoro pentanol, 4,4,5,5,5-pentafluoro-pentanol, 4,4,5,5,5-pentafluorpentan-1-ol | |
| 4,4,5,5,5-pentafluoropentan-1-ol |
Safety and Handling
EINECSNumber : (2)-3955
TSCA : No