missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4',4″-Tris(diphenylamino)triphenylamine 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 247°C |
| Color | Yellow |
| Quantity | 1 g |
| Formula Weight | 746.96 |
| Percent Purity | ≥97.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4,4′,4′′-Tris(diphenylamino)triphenylamine |
Safety and Handling
| TSCA | No |