accessibility menu, dialog, popup

UserName

CAS RN 105389-36-4

NNNN

CAS RN 105389-36-4

IUPAC Name: N1,N1-bis[4-(diphenylamino)phenyl]-N4,N4-diphenylbenzene-1,4-diamine
Synonyms: N1,N1-bis[4-(diphenylamino)phenyl]-N4,N4-diphenylbenzene-1,4-diamine
Molecular Weight (g/mol): 746.96
Molecular Formula: C54H42N4
InChi Key: IYZMXHQDXZKNCY-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=C1)N(C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results