missing translation for 'onlineSavingsMsg'
Learn More

4,4',4″-Tris(diphenylamino)triphenylamine 97.0+%, TCI America™

Catalog No. T3062200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3062200MG 200 mg
T30621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3062200MG Supplier TCI America Supplier No. T3062200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 105389-36-4
Molecular Formula C54H42N4
Molecular Weight (g/mol) 746.958
MDL Number MFCD01310677
InChI Key IYZMXHQDXZKNCY-UHFFFAOYSA-N
Synonym TDATA
PubChem CID 10952754
IUPAC Name 1-N,1-N-diphenyl-4-N,4-N-bis[4-(N-phenylanilino)phenyl]benzene-1,4-diamine
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)N(C4=CC=C(C=C4)N(C5=CC=CC=C5)C6=CC=CC=C6)C7=CC=C(C=C7)N(C8=CC=CC=C8)C9=CC=CC=C9

Specifications

Melting Point 247°C
Color Yellow
Quantity 200 mg
Formula Weight 746.96
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′,4′′-Tris(diphenylamino)triphenylamine