missing translation for 'onlineSavingsMsg'
Learn More

4,4',4″-Trimethyltrityl alcohol, 98+%

Catalog No. p-7043732
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0239206 5 g
AAL0239214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0239206 Supplier Thermo Scientific Chemicals Supplier No. L0239206

CAS: 3247-00-5 | C22H22O | 302.417 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3247-00-5
Molecular Formula C22H22O
Molecular Weight (g/mol) 302.417
MDL Number MFCD00014919
InChI Key DNWQXZDDISHGRM-UHFFFAOYSA-N
Synonym 4,4',4-trimethyltrityl alcohol, tris 4-methylphenyl methanol, methanol, tri-p-tolyl, tri-p-tolylmethanol, tri p-tolyl methanol, 4,4'-dimethyl-4-methyltrityl alcohol, benzenemethanol, 4-methyl-.alpha.,.alpha.-bis 4-methylphenyl, acmc-20amt7, tris 4-methylphenyl methan-1-ol, 4,4-trimethyltriphenylmethanol
PubChem CID 76733
IUPAC Name tris(4-methylphenyl)methanol
SMILES CC1=CC=C(C=C1)C(C2=CC=C(C=C2)C)(C3=CC=C(C=C3)C)O

Specifications

Melting Point 92°C to 96°C
Quantity 5 g
Beilstein 2055803
Formula Weight 302.42
Percent Purity ≥98%
Chemical Name or Material 4,4',4''-Trimethyltrityl alcohol
EINECSNumber 221-822-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only