missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluorocrotononitrile, 96%

Catalog No. aal1687003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1687003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1687003 Supplier Thermo Scientific Chemicals Supplier No. L1687003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 406-86-0 | C4H2F3N | 121.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 406-86-0
Molecular Formula C4H2F3N
Molecular Weight (g/mol) 121.06
MDL Number MFCD00190645
InChI Key LHWSEFCIRYVTLZ-UHFFFAOYSA-N
Synonym 4,4,4-trifluorocrotononitrile, 4,4,4-trifluorocrotonitrile, 2e-4,4,4-trifluorobut-2-enenitrile, 3-trifluoromethylacrylonitrile, 4,4,4-trifluorobut-2-enenitrile, 4,4,4-trifluoro-2-butenenitrile, trifluorocrotononitrile, ka, a`\d`dadadtarjfppp, e-4,4,4-trifluoro-2-butenenitrile, e-4,4,4-trifluorobut-2-enenitrile
PubChem CID 5708782
IUPAC Name (E)-4,4,4-trifluorobut-2-enenitrile
SMILES FC(F)(F)C=CC#N

Specifications

Boiling Point 84°C to 85°C
Quantity 1 g
UN Number UN3276
Beilstein 6381034
Formula Weight 121.06
Percent Purity 96%
Chemical Name or Material 4,4,4-Trifluorocrotononitrile
Hazard Category H301+H311-H315-H319-H330-H335
Hazard Statement GHS H Statement
H300-H310-H330-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only