CAS RN 406-86-0
IUPAC Name:
4,4,4-trifluorobut-2-enenitrile
Synonyms:
4,4,4-trifluorobut-2-enenitrile
Molecular Weight (g/mol):
121.06
Molecular Formula:
C4H2F3N
InChi Key:
LHWSEFCIRYVTLZ-UHFFFAOYSA-N
SMILES:
FC(F)(F)C=CC#N
1
–
1
of
1
results
1
Promotions Available
4,4,4-Trifluorocrotononitrile, 96%
4,4,4-Trifluorocrotononitrile, 96%
CAS: 406-86-0 Molecular Formula: C4H2F3N Molecular Weight (g/mol): 121.06 MDL Number: MFCD00190645 InChI Key: LHWSEFCIRYVTLZ-UHFFFAOYSA-N Synonym: 4,4,4-trifluorocrotononitrile,4,4,4-trifluorocrotonitrile,2e-4,4,4-trifluorobut-2-enenitrile,3-trifluoromethylacrylonitrile,4,4,4-trifluorobut-2-enenitrile,4,4,4-trifluoro-2-butenenitrile,trifluorocrotononitrile,ka,a`\d`dadadtarjfppp,e-4,4,4-trifluoro-2-butenenitrile,e-4,4,4-trifluorobut-2-enenitrile PubChem CID: 5708782 IUPAC Name: (E)-4,4,4-trifluorobut-2-enenitrile SMILES: FC(F)(F)C=CC#N