missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluoro-3-hydroxy-3-methylbutyric acid, 98%

Catalog No. AAL1687306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1687306 5 g
AAL1687303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1687306 Supplier Thermo Scientific Chemicals Supplier No. L1687306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 338-03-4 | C5H7F3O3 | 172.103 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 338-03-4
Molecular Formula C5H7F3O3
Molecular Weight (g/mol) 172.103
MDL Number MFCD00039531
InChI Key JCJGOWYCFRASTM-UHFFFAOYSA-N
Synonym 3-trifluoromethyl-3-hydroxybutyric acid, 4,4,4-trifluoro-3-hydroxy-3-methylbutyric acid, 3-hydroxy-3-methyl-4,4,4-trifluorobutyric acid, 4,4,4-trifluoro-3-hydroxy-3-methyl-butyric acid, butanoic acid, 4,4,4-trifluoro-3-hydroxy-3-methyl, pubchem12644, acmc-20an2r, 3-hydroxy-3-trifluoromethylbutyric acid, 3-hydroxy-3-trifluoromethyl butyric acid
PubChem CID 136168
IUPAC Name 4,4,4-trifluoro-3-hydroxy-3-methylbutanoic acid
SMILES CC(CC(=O)O)(C(F)(F)F)O

Specifications

Melting Point 39°C to 41°C
Boiling Point 87°C to 90°C (0.5 mmHg)
Refractive Index 1.388
Quantity 5 g
UN Number UN3261
Formula Weight 172.1
Percent Purity 98%
Chemical Name or Material 4,4,4-Trifluoro-3-hydroxy-3-methylbutyric acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only