accessibility menu, dialog, popup

UserName

CH3(S)HOOOFFF

CAS RN 338-03-4

IUPAC Name: (3S)-4,4,4-trifluoro-3-hydroxy-3-methylbutanoate
Synonyms: (3S)-4,4,4-trifluoro-3-hydroxy-3-methylbutanoate
Molecular Weight (g/mol): 171.1
Molecular Formula: C5H6F3O3
InChi Key: JCJGOWYCFRASTM-BYPYZUCNSA-M
SMILES: C[C@](O)(CC([O-])=O)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)