missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluoro-1-butanol 98.0+%, TCI America™

Catalog No. p-7137414
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
T27811G 1 g
T27815G 5 g
2 options

Catalog No. T27811G

Supplier: TCI America T27811G

Only null left

Chemical Identifiers

CAS 461-18-7
Molecular Formula C4H7F3O
Molecular Weight (g/mol) 128.094
MDL Number MFCD00041483
InChI Key VKRFUGHXKNNIJO-UHFFFAOYSA-N
Synonym 4,4,4-trifluoro-1-butanol, 4,4,4-trifluorobutanol, 4,4,4-trifluorobutanol-1, 4,4,4-trifluoro-butan-1-ol, 1-butanol, 4,4,4-trifluoro, pubchem15240, dl-bicarnesine hydrochloride, ksc237k9h, 4,4,4-trifluoro-1-butanol #, 4,4,4-tris fluoranyl butan-1-ol
PubChem CID 136307
IUPAC Name 4,4,4-trifluorobutan-1-ol
SMILES C(CC(F)(F)F)CO

Specifications

Color Yellow
Boiling Point 125°C
Quantity 1 g
UN Number 1987
Formula Weight 128.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4,4,4-Trifluoro-1-butanol